Skip to Content
Merck
All Photos(1)

Key Documents

84010

Sigma-Aldrich

Rubidium nitrate

purum p.a., ≥99.0% (T)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
RbNO3
CAS Number:
Molecular Weight:
147.47
EC Number:
MDL number:
UNSPSC Code:
12352302

grade

purum p.a.

Quality Level

Assay

≥99.0% (T)

density

3.11 g/mL at 25 °C (lit.)

anion traces

chloride (Cl-): ≤100 mg/kg
sulfate (SO42-): ≤100 mg/kg

cation traces

Ca: ≤50 mg/kg
Cd: ≤50 mg/kg
Co: ≤50 mg/kg
Cu: ≤50 mg/kg
Fe: ≤50 mg/kg
K: ≤5000 mg/kg
Na: ≤100 mg/kg
Ni: ≤50 mg/kg
Pb: ≤50 mg/kg
Zn: ≤50 mg/kg

SMILES string

[Rb+].[O-][N+]([O-])=O

InChI

1S/NO3.Rb/c2-1(3)4;/q-1;+1

InChI key

RTHYXYOJKHGZJT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service