Skip to Content
Merck
All Photos(1)

Key Documents

81338

Sigma-Aldrich

Poly-L-lysine hydrobromide

mol wt 30,000-70,000

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352200

form

fibers

mol wt

30,000-70,000

storage temp.

−20°C

SMILES string

Cl.NCCCCC(N)C(O)=O

InChI

1S/C18H38N6O4/c19-10-4-1-7-13(22)16(25)23-14(8-2-5-11-20)17(26)24-15(18(27)28)9-3-6-12-21/h13-15H,1-12,19-22H2,(H,23,25)(H,24,26)(H,27,28)/t13-,14-,15-/m0/s1

InChI key

WBSCNDJQPKSPII-KKUMJFAQSA-N

Looking for similar products? Visit Product Comparison Guide

Components

Poly-L-lysine is a positively charged amino acid polymer with approximately one HBr per lysine residue. The hydrobromide allows the poly-L-lysine to be in a crystalline form soluble in water. A small amount of product may be found in the beta structure because the HBr interferes with hydrogen bonding between amino and either the carboxyl groups or N or O containing moieties.

Caution

Sterile solutions are stable for up to 2 years when stored at 2-8°C. It should be stored desiccated at -20°C.

Analysis Note

Molecular weight based on viscosity. Also assayed by MALLS.

Other Notes

For additional technical information on polyamino acids please visit the Polyamino acid FAQ resource.
A polycation which binds to DNA, red cell membrane and any negatively charged protein. For the preparation of polycationic beads, useful in immobilization techniques.; Immobilization of membranes; Immobilization of plant protoplasts; In cell culture, immobilization by microencapsulation with the alginate-poly- L-lysine microcapsule system; Drug-Polylysine conjugates: Their potential for chemotherapy and for the study of endocytosis

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Advances in the Biosciences, 61, 3-3 (1977)
G Payares et al.
Molecular and biochemical parasitology, 23(2), 129-137 (1987-03-01)
Poly-lysine coated beads attached readily onto Schistosoma mansoni. On detachment, the beads removed membranes from the surface of the tegument. Analysis of the proteins of the detached membranes showed that three major proteins of 94, 73 and 62 kDa were
G.A. King et al.
Biotechnology Progress, 3, 231-231 (1987)
H.J.-P. Ryser et al.
NATO ASI Ser; Ser. A, 113, 103-103 (1986)
R. Vankova et al.
Physiologia Plantarum, 70, 1-1 (1987)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service