Skip to Content
Merck
All Photos(1)

Documents

78975

Supelco

α-Methylbenzyl acetate

analytical standard

Synonym(s):

α-Methylbenzyl acetate, α-Methylbenzenemethanol 1-acetate, 1-Phenylethyl acetate, Gardenol, Styrallyl acetate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CO2CH(CH3)C6H5
CAS Number:
Molecular Weight:
164.20
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥99.0% (GC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.493-1.497
n20/D 1.494 (lit.)

bp

94-95 °C/12 mmHg (lit.)

density

1.028 g/mL at 25 °C (lit.)

application(s)

cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care

format

neat

SMILES string

CC(OC(C)=O)c1ccccc1

InChI

1S/C10H12O2/c1-8(12-9(2)11)10-6-4-3-5-7-10/h3-8H,1-2H3

InChI key

QUMXDOLUJCHOAY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

198.5 °F

Flash Point(C)

92.5 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Guzman Torrelo et al.
Chembiochem : a European journal of chemical biology, 16(4), 625-630 (2015-02-17)
In 2009, we reported that the product of the gene SCJ21.16 (XFa0032) from Xylella fastidiosa, a xylem-restricted plant pathogen that causes a range of diseases in several important crops, encodes a protein (XfHNL) with putative hydroxynitrile lyase activity. Sequence analysis

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service