Skip to Content
Merck
All Photos(1)

Key Documents

73105

Supelco

Glyceryl tributyrate

analytical standard

Synonym(s):

1,2,3-Tributyrylglycerol, Glycerol tributyrate, Tributyrin

Sign Into View Organizational & Contract Pricing

Select a Size

1 ML
€56.70

€56.70


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
1 ML
€56.70

About This Item

Linear Formula:
(CH3CH2CH2COOCH2)2CHOCOCH2CH2CH3
CAS Number:
Molecular Weight:
302.36
Beilstein:
1714746
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

€56.70


Please contact Customer Service for Availability

Request a Bulk Order

grade

analytical standard

Quality Level

Assay

≥98.0% (GC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.434-1.436
n20/D 1.435 (lit.)

bp

129-131 °C/0.03 mmHg (lit.)
287-288 °C (lit.)

density

1.032 g/mL at 20 °C (lit.)

application(s)

food and beverages

format

neat

SMILES string

CCCC(=O)OCC(COC(=O)CCC)OC(=O)CCC

InChI

1S/C15H26O6/c1-4-7-13(16)19-10-12(21-15(18)9-6-3)11-20-14(17)8-5-2/h12H,4-11H2,1-3H3

InChI key

UYXTWWCETRIEDR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

345.2 °F - closed cup

Flash Point(C)

174 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Gail A Cresci et al.
Alcoholism, clinical and experimental research, 38(6), 1489-1501 (2014-06-04)
Excessive alcohol consumption leads to liver disease. Interorgan crosstalk contributes to ethanol (EtOH)-induced liver injury. EtOH exposure causes gut dysbiosis resulting in negative alterations in intestinal fermentation byproducts, particularly decreased luminal butyrate concentrations. Therefore, in the present work, we investigated
Soon-Seok Hong et al.
International journal of pharmaceutics, 483(1-2), 142-150 (2015-02-11)
A high drug-loading capacity is a critical factor for the clinical development of liposomal formulations. The accommodation of hydrophobic drugs within the liposomal membrane is often limited in saturated phosphatidylcholine (PC)-based liposomes owing to the rigidity of the lipid acyl
Mette U Anby et al.
Molecular pharmaceutics, 11(11), 4069-4083 (2014-09-30)
The impact of gastrointestinal (GI) processing and first pass metabolism on danazol oral bioavailability (BA) was evaluated after administration of self-emulsifying drug delivery systems (SEDDS) in the rat. Danazol absolute BA was determined following oral and intraduodenal (ID) administration of

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service