Skip to Content
Merck
All Photos(1)

Key Documents

69692

Sigma-Aldrich

3-Buten-2-one

purum, ≥95.0% (GC)

Synonym(s):

Methyl vinyl ketone, Vinyl methyl ketone

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH2=CHCOCH3
CAS Number:
Molecular Weight:
70.09
Beilstein:
506021
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
Pricing and availability is not currently available.

vapor density

1.3 (vs air)

Quality Level

vapor pressure

310 mmHg ( 55 °C)
71 mmHg ( 20 °C)

grade

purum

Assay

≥95.0% (GC)

form

liquid

contains

~0.5% acetic acid as stabilizer
~0.5% hydroquinone as stabilizer

expl. lim.

15.64 %

impurities

~5% water

refractive index

n20/D 1.411 (lit.)
n20/D 1.412

bp

81 °C (lit.)

density

0.86 g/mL at 20 °C
0.864 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

CC(=O)C=C

InChI

1S/C4H6O/c1-3-4(2)5/h3H,1H2,2H3

InChI key

FUSUHKVFWTUUBE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Caution

may discolor to brown on storage

Signal Word

Danger

Hazard Classifications

Acute Tox. 1 Dermal - Acute Tox. 1 Inhalation - Acute Tox. 2 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Flam. Liq. 2 - Skin Corr. 1B - Skin Sens. 1

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

19.4 °F - closed cup

Flash Point(C)

-7 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yonggui Chi et al.
Journal of the American Chemical Society, 130(20), 6322-6323 (2008-04-25)
Non-interpenetrating star polymer catalysts designed to mimic the site isolation characteristics of enzymes enable the one-pot combination of multiple otherwise incompatible catalysts for asymmetric cascade reactions that involve iminium, enamine, and H-bonding catalysis. Control experiments replacing star polymer catalysts with
Hyun Joo Lee et al.
Molecules (Basel, Switzerland), 17(6), 7523-7532 (2012-06-20)
The enantioselective conjugate addition reaction of 3-aryl-substituted oxindoles with methyl vinyl ketone promoted by binaphthyl-modified bifunctional organocatalysts was investigated. The corresponding Michael adducts, containing a quaternary center at the C3-position of the oxindoles, were generally obtained in high yields with
Quan Cai et al.
Organic letters, 14(12), 3040-3043 (2012-05-26)
A formal [4 + 2] cycloaddition of 2,3-disubstituted indoles with vinyl methyl ketone was realized in the presence of a catalytic amount of quinine-derived primary amine and pentafluorobenzoic acid. This method provides bridged-ring indoline scaffolds containing two quaternary carbon centers
Grant R Krow et al.
The Journal of organic chemistry, 72(9), 3458-3466 (2007-03-31)
Diene substituent effects on the regiochemical and stereochemical outcomes of uncatalyzed Diels-Alder reactions of N-alkoxycarbonyl-1,2-dihydropyridines with both styrene and methyl vinyl ketone (MVK) were studied. Alkyl substitution on the diene in all cases examined resulted in a kinetic preference for
Palladium-mediated annulation of vinyl aziridines with Michael acceptors: stereocontrolled synthesis of substituted pyrrolidines and its application in a formal synthesis of (-)-α-kainic acid.
Martin A Lowe et al.
Angewandte Chemie (International ed. in English), 50(28), 6370-6374 (2011-06-01)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service