Skip to Content
Merck
All Photos(1)

Documents

6900000140BC

Millipore

BIOHAZARD WASTE BAGS (BOX OF 10)

BioControl Systems, for use with Gemini®

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41122409
NACRES:
NA.84

packaging

pkg of 10 units

manufacturer/tradename

BioControl Systems
Gemini®

technique(s)

ELISA: suitable
enzyme immunoassay: suitable

application(s)

food and beverages

compatibility

for use with Gemini®

storage temp.

room temp

Legal Information

Gemini is a registered trademark of Stratec

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service