Skip to Content
Merck
All Photos(1)

Documents

63973

Millipore

M-17 Broth without lactose

suitable for microbiology, NutriSelect® Basic

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41171607
NACRES:
NA.74

form

solid

Quality Level

shelf life

limited shelf life, expiry date on the label

composition

ascorbic acid, 0.5 g/L
magnesium sulfate, 0.25 g/L
meat extract, 5 g/L
meat peptone (peptic), 2.5 g/L
sodium glycerophosphate, 19 g/L
soya peptone (papainic), 5 g/L
yeast extract, 2.5 g/L

manufacturer/tradename

NutriSelect® Basic

pH

7.0±0.2 (25 °C)

application(s)

microbiology

Application

With the addition of lactose this medium is recommended to use for the cultivation of Streptococcus thermophilus.

Preparation Note

Directions:
Suspend 37 g of powder in 1 litre of distilled water and let it soak. Heat to boiling and dispense into suitable containers. Sterilize by autoclaving at 121°C for 15 minutes.

Footnote

We offer two media types: the superior granulated GranuCult® and the cost-efficient powdered NutriSelect® culture media, depending on your needs.
The designations basic, plus, or prime are added to indicate the quality control level, from basic quality control to standard QC plus to prime for full regulatory compliance.

Legal Information

GRANUCULT is a registered trademark of Merck KGaA, Darmstadt, Germany
NutriSelect is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service