Skip to Content
Merck
All Photos(1)

Key Documents

56912

Supelco

Aluminum bronze

manufactured by MBH Analytical Ltd (32X ALB2)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

material

disks

grade

analytical standard

technique(s)

XRF: suitable

diam. × H

43 mm × 20 mm , dimensions are approximate

format

matrix material

Analysis Note

Values of analytes vary lot to lot.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

No data available

Flash Point(C)

No data available


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service