Skip to Content
Merck
All Photos(1)

Key Documents

55398

Supelco

L-2-Aminoadipic acid

analytical standard

Synonym(s):

(S)-2-Aminohexanedioic acid, L-Homoglutamic acid, Aad

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HO2C(CH2)3CH(NH2)CO2H
CAS Number:
Molecular Weight:
161.16
Beilstein:
1724348
MDL number:
UNSPSC Code:
41116107

grade

analytical standard

Assay

≥98% (TLC)

optical activity

[α]25/D 24±1°, c = 2 in 5 M HCl

shelf life

limited shelf life, expiry date on the label

mp

203-205 °C (dec.) (lit.)

application(s)

clinical testing

format

neat

SMILES string

N[C@@H](CCCC(O)=O)C(O)=O

InChI

1S/C6H11NO4/c7-4(6(10)11)2-1-3-5(8)9/h4H,1-3,7H2,(H,8,9)(H,10,11)/t4-/m0/s1

InChI key

OYIFNHCXNCRBQI-BYPYZUCNSA-N

Looking for similar products? Visit Product Comparison Guide

General description

L-2-Aminoadipic acid is the six-carbon homolog of glutamate and a metabolite of L-lysine.

Application

L-2-Aminoadipic acid may be used as an analytical standard in the following:
  • For the quantification of 2-aminoadipic acid in acid hydrolysates of processed skin samples using gas chromatography coupled to mass spectrometry (GC-MS).
  • To investigate its effect on proteolysis and protein synthesis in C2C12 myotubes.

It may be used as an internal standard for the determination of free amino acids and ammonium ion contents in Crocus sativus L. from different geographical origins using high-performance liquid chromatography with diode array detection (HPLC-DAD).

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Determination of free amino acids and ammonium ion in saffron (Crocus sativus L.) from different geographical origins
Campo dPC, et al.
Food Chemistry, 114(4), 1542-1548 (2009)
Regulatory effects of the L-lysine metabolites, L-2-aminoadipic acid and L-pipecolic acid, on protein turnover in C2C12 myotubes
Sato T, et al.
Bioscience, Biotechnology, and Biochemistry, 80(11), 2168-2175 (2016)
David R Sell et al.
The Biochemical journal, 404(2), 269-277 (2007-02-23)
We hypothesized that the epsilon-amino group of lysine residues in longlived proteins oxidatively deaminates with age forming the carbonyl compound, allysine (alpha-aminoadipic acid-delta-semialdehyde), which can further oxidize into 2-aminoadipic acid. In the present study, we measured both products in insoluble
Maho Shibata et al.
Investigative ophthalmology & visual science, 53(7), 3726-3732 (2012-05-17)
To investigate the involvement of glial cells in the autoregulation of optic nerve head (ONH) blood flow in response to elevated intraocular pressure (IOP). Rabbit eyes were treated with an intravitreal injection of l-2-aminoadipic acid (LAA), a gliotoxic compound. Twenty-four

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service