Skip to Content
Merck
All Photos(1)

Key Documents

53162

Supelco

Laboratory pH-Electrode

Glass body (Double junction refillable)

Synonym(s):

pH-Electrode for the laboratory

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
26111700
NACRES:
NB.61

material

glass shaft

grade

for ion-selective electrodes

temperature measuring range

0-100 °C

pH

0-14
7.0±0.25(internal buffer)

General description

Electrolyte refillable, KCl c= 3 mol/L, Ag+-free
S7 connector
Visit our Sensor Applications portal to learn more.

Application

For reliabel pH measurement in the water industry and for process chemistry.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service