Skip to Content
Merck
All Photos(2)

Key Documents

51825

Supelco

Bemotrizinol

analytical standard

Synonym(s):

2,4-Bis[4-(2-ethylhexyloxy)-2-hydroxyphenyl]-6-(4-methoxyphenyl)-1,3,5-triazine

Sign Into View Organizational & Contract Pricing

Select a Size

25 MG
€176.00

€176.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
25 MG
€176.00

About This Item

Empirical Formula (Hill Notation):
C38H49N3O5
CAS Number:
Molecular Weight:
627.81
Beilstein:
11322254
MDL number:
UNSPSC Code:
77101502
PubChem Substance ID:
NACRES:
NA.24

€176.00


Please contact Customer Service for Availability

Request a Bulk Order

grade

analytical standard

Quality Level

Assay

≥98.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

composition

carbon content, 72.70% (theory)
hydrogen content, 7.87% (theory)
nitrogen content, 6.69% (theory)

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

mp

77-85 °C

application(s)

cleaning products
cosmetics
environmental
food and beverages
personal care

format

neat

SMILES string

CCCCC(CC)COc1ccc(c(O)c1)-c2nc(nc(n2)-c3ccc(OCC(CC)CCCC)cc3O)-c4ccc(OC)cc4

InChI

1S/C38H49N3O5/c1-6-10-12-26(8-3)24-45-30-18-20-32(34(42)22-30)37-39-36(28-14-16-29(44-5)17-15-28)40-38(41-37)33-21-19-31(23-35(33)43)46-25-27(9-4)13-11-7-2/h14-23,26-27,42-43H,6-13,24-25H2,1-5H3

InChI key

XVAMCHGMPYWHNL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Bemotrizinol is a fat-soluble organic UV filter that belongs to the family of methoxybenzenes. It is commonly used as a constituent of sunscreen products due to its ability to absorb UV-A radiations. It is commonly sold as Tinosorb S and Escalol S.[1]

Application

The analytical standard can also be used as follows:
  • Multi-residue analysis of five UV filters in sunscreen lotion and human outermost skin samples by ultrasound-vortex-assisted DLLME (US-VA-DLLME) and high-performance liquid chromatography-diode array detection (HPLC-DAD)
  • Quantification of nine organic UV filters in coastal sand samples using an optimized high-performance liquid chromatography (HPLC) method based on a biphenyl core-shell column, in combination with UV and fluorescence detection
  • Determination of the analyte in topical pharmaceutical formulations by reversed-phase high-performance liquid chromatography method (RP-HPLC).[1]

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Other Notes

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Development and validation of a stability indicating RP-HPLC method for the determination of two sun protection factors (Koptrizon and Tinosorb S) in topical pharmaceutical formulations using experimental designs
Roy C and Chakrabarty J
ISRN Chromatography, 2013 (2013)
Diogo Noin de Oliveira et al.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences, 988, 13-19 (2015-03-07)
Sunlight exposure causes several types of injury to humans, especially on the skin; among the most common harmful effects due to ultraviolet (UV) exposure are erythema, pigmentation and lesions in DNA, which may lead to cancer. These long-term effects are

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service