Skip to Content
Merck
All Photos(1)

Documents

42551

Sigma-Aldrich

1,2-Dipalmitoyl-rac-glycero-3-phosphoethanolamine

≥99.0% (TLC)

Synonym(s):

1,2-Dihexadecanoyl-rac-glycero-3-phosphoethanolamine, rac-Phosphatidylethanolamine, 1,2-dipalmitoyl, DL-β,γ-Dipalmitoyl-α-cephalin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C37H74NO8P
CAS Number:
Molecular Weight:
691.96
Beilstein:
1730602
EC Number:
MDL number:
UNSPSC Code:
12352211

Assay

≥99.0% (TLC)

storage temp.

−20°C

SMILES string

CCCCCCCCCCCCCCCC(=O)OCC(COP(O)(=O)OCCN)OC(=O)CCCCCCCCCCCCCCC

InChI

1S/C37H74NO8P/c1-3-5-7-9-11-13-15-17-19-21-23-25-27-29-36(39)43-33-35(34-45-47(41,42)44-32-31-38)46-37(40)30-28-26-24-22-20-18-16-14-12-10-8-6-4-2/h35H,3-34,38H2,1-2H3,(H,41,42)

InChI key

SLKDGVPOSSLUAI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

replaced by


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service