Skip to Content
Merck
All Photos(2)

Key Documents

35344

Supelco

Albendazole-(methyl-d3)

VETRANAL®, analytical standard

Synonym(s):

Methyl-d3 5-(propylthio)-2-benzimidazolecarbamate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12D3H12N3O2S
CAS Number:
Molecular Weight:
268.35
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

VETRANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

forensics and toxicology
pharmaceutical (small molecule)

format

neat

mass shift

M+3

SMILES string

[2H]C([2H])([2H])OC(=O)Nc1nc2cc(SCCC)ccc2[nH]1

InChI

1S/C12H15N3O2S/c1-3-6-18-8-4-5-9-10(7-8)14-11(13-9)15-12(16)17-2/h4-5,7H,3,6H2,1-2H3,(H2,13,14,15,16)/i2D3

InChI key

HXHWSAZORRCQMX-BMSJAHLVSA-N

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

VETRANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazardEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Repr. 2 - STOT RE 2 Oral

Target Organs

Adrenal gland,Blood,male reproductive organs,spleen

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Michael Walker et al.
Food additives & contaminants. Part A, Chemistry, analysis, control, exposure & risk assessment, 34(4), 489-493 (2016-10-04)
Albendazole, one of the benzimidazole anthelmintics, is used in ruminants and has maximum residue limits in muscle, fat and other tissue owing to reported teratogenicity. Albendazole is extensively metabolised in domestic animals and humans with rapid conversion to a sulphoxide

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service