Skip to Content
Merck
All Photos(1)

Key Documents

33983

Supelco

Dibutyl succinate

PESTANAL®, analytical standard

Synonym(s):

Tabutrex

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)3OCOCH2CH2COO(CH2)3CH3
CAS Number:
Molecular Weight:
230.30
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care

format

neat

SMILES string

CCCCOC(=O)CCC(=O)OCCCC

InChI

1S/C12H22O4/c1-3-5-9-15-11(13)7-8-12(14)16-10-6-4-2/h3-10H2,1-2H3

InChI key

YUXIBTJKHLUKBD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Dibutyl succinate is an insect repellent, which is approved for use on farm animals and livestock.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

275.0 °F - open cup

Flash Point(C)

135 °C - open cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chemistry of Pesticides. (2012)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service