Skip to Content
Merck
All Photos(1)

Key Documents

320544

Sigma-Aldrich

Thionyl chloride

ReagentPlus®, ≥99%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
SOCl2
CAS Number:
Molecular Weight:
118.97
Beilstein:
1209273
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:

vapor pressure

97 mmHg ( 20 °C)

product line

ReagentPlus®

Assay

≥99%

form

liquid

refractive index

n20/D 1.518 (lit.)

bp

79 °C (lit.)

mp

−105 °C (lit.)

density

1.631 g/mL at 25 °C (lit.)

SMILES string

ClS(Cl)=O

InChI

1S/Cl2OS/c1-4(2)3

InChI key

FYSNRJHAOHDILO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

ReagentPlus is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Skull and crossbonesCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Eye Dam. 1 - Skin Corr. 1A - STOT SE 3

Target Organs

Respiratory system

Supplementary Hazards

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Meng Zhou et al.
Molecules (Basel, Switzerland), 20(6), 10122-10130 (2015-06-04)
Pincer (Phebox)Ir(mesityl)(OAc) (2) (Phebox = 3,5-dimethylphenyl-2,6-bis(oxazolinyl)) complex, formed by benzylic C-H activation of mesitylene (1,3,5-trimethylbenzene) using (Phebox)Ir(OAc)2OH2 (1), was treated with thionyl chloride to rapidly form 1-(chloromethyl)-3,5-dimethylbenzene in 50% yield at 23 °C. A green species was obtained at the
Eagleson M.
Concise Encyclopedia Chemistry, 1056-1056 (1994)
Eagleson M.
Concise Encyclopedia Chemistry, 118-118 (1994)
Eagleson M.
Concise Encyclopedia Chemistry, 1049-1049 (1994)
Feng Xu et al.
The Journal of organic chemistry, 73(1), 312-315 (2007-12-07)
A simple, one-pot preparation of cyclic amines via efficient chlorination of amino alcohols with use of SOCl(2) has been developed. This approach obviates the need for the classical N-protection/O-activation/cyclization/deprotection sequence commonly employed for this type of transformation. The reaction pathways

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service