28614-U
Hamilton® 701N syringe
701N (Slimline), volume 10 μL, needle size 26s ga (bevel tip), needle L 51 mm (2 in.)
About This Item
Recommended Products
material
bevel tip stainless steel (point style 2)
needle size
26s ga (bevel tip)
feature
Microliter™
Syringe, Fixed Needle
packaging
pkg of 1 ea
manufacturer/tradename
Hamilton 203072
needle L
51 mm (2 in.)
volume
10 μL
compatibility
for use with CTC A200S PAL Instruments (cemented needle)
Looking for similar products? Visit Product Comparison Guide
Related Categories
Legal Information
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service