Skip to Content
Merck
All Photos(1)

Documents

28314

Sigma-Aldrich

2-Cyanoethyl N,N,N′,N′-tetraisopropylphosphorodiamidite

purum, ≥98.0% (31P-NMR)

Synonym(s):

Bis(diisopropylamino)(2-cyanoethoxy)phosphine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
{[(CH3)2CH]2N}2POCH2CH2CN
CAS Number:
Molecular Weight:
301.41
Beilstein:
3590103
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

grade

purum

Assay

≥98.0% (31P-NMR)

refractive index

n20/D 1.470 (lit.)
n20/D 1.470

bp

100 °C/0.5 mmHg (lit.)

density

0.949 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

CC(C)N(C(C)C)P(OCCC#N)N(C(C)C)C(C)C

InChI

1S/C15H32N3OP/c1-12(2)17(13(3)4)20(19-11-9-10-16)18(14(5)6)15(7)8/h12-15H,9,11H2,1-8H3

InChI key

RKVHNYJPIXOHRW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Other Notes

Phosphitylation reagent used in nucleotide synthesis by the phosphite method ; Phosphitylation of other compounds

replaced by

Product No.
Description
Pricing

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 3 - Flam. Liq. 3 - Skin Sens. 1B

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

119.3 °F - closed cup

Flash Point(C)

48.5 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

W. Bannwarth et al.
Helvetica Chimica Acta, 71, 1517-1517 (1988)
W. Bannwarth et al.
Helvetica Chimica Acta, 70, 175-175 (1987)
Y. Jl et al.
Tetrahedron, 46, 487-487 (1990)
M Morillo et al.
Lipids, 31(5), 541-546 (1996-05-01)
A simple chemical method for the synthesis of 1,2-diacyl-sn-glycerophosphatidylserine (PS), with the same fatty acid composition in the sn-1 and sn-2 glycerol positions as egg phosphatidylcholine (PC), is described. PS synthesis was carried out by a phosphite-triester approach, using 2-cyanoethyl-N,N,N',N'-tetraisopropylphosphorodiamidite
Improved synthesis of (Pri2 N)2POCH2CH2CN.
J Nielsen et al.
Nucleic acids research, 15(8), 3626-3626 (1987-04-24)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service