23590
Sodium 4-chlorobenzenesulfinate hydrate
technical, ≥90% (HPLC)
Synonym(s):
4-Chlorobenzenesulfinic acid sodium salt hydrate
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C6H4ClNaO2S · xH2O
CAS Number:
Molecular Weight:
198.60 (anhydrous basis)
Beilstein:
3575604
EC Number:
MDL number:
UNSPSC Code:
12352103
grade
technical
Assay
≥90% (HPLC)
impurities
≤40% water
SMILES string
[Na+].[O-]S(=O)c1ccc(Cl)cc1
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service