Skip to Content
Merck
All Photos(1)

Documents

21345

Sigma-Aldrich

(−)-Camphoric acid

purum, unnatural form, ≥99.0% (T)

Synonym(s):

(1S,3R)-1,2,2-Trimethyl-1,3-cyclopentanedicarboxylic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H16O4
CAS Number:
Molecular Weight:
200.23
Beilstein:
1874674
MDL number:
UNSPSC Code:
51113400
PubChem Substance ID:

grade

purum

Assay

≥99.0% (T)

form

solid

optical activity

[α]20/D −46±2°, c = 1% in ethanol

quality

unnatural form

mp

184-189 °C

SMILES string

CC1(C)[C@@H](CC[C@]1(C)C(O)=O)C(O)=O

InChI

1S/C10H16O4/c1-9(2)6(7(11)12)4-5-10(9,3)8(13)14/h6H,4-5H2,1-3H3,(H,11,12)(H,13,14)/t6-,10+/m0/s1

InChI key

LSPHULWDVZXLIL-QUBYGPBYSA-N

replaced by

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Uri Hananel et al.
Proceedings of the National Academy of Sciences of the United States of America, 116(23), 11159-11164 (2019-05-18)
Symmetry plays a crucial part in our understanding of the natural world. Mirror symmetry breaking is of special interest as it is related to life as we know it. Studying systems which display chiral amplification, therefore, could further our understanding
Karina Petersen et al.
Aquatic toxicology (Amsterdam, Netherlands), 190, 150-161 (2017-07-18)
Many environmental matrices contaminated with organic pollutants derived from crude oil or degraded petroleum contain mixtures so complex that they are typically unresolved by conventional analytical techniques such as gas chromatography. The resulting chromatographic features have become known as 'humps'
Xuan Wang et al.
Electrophoresis, 39(17), 2236-2245 (2018-05-26)
In this study, a metal-organic framework (MOF), [Mn(cam)(bpy)], was synthesized and characterized by thermogravimetric analysis, scanning electron microscopy, and Fourier transform infrared spectrometry. An open-tubular capillary column was fabricated from [Mn(cam)(bpy)] via the amide coupling method. Ten types of sulfonamides

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service