Skip to Content
Merck
All Photos(1)

Key Documents

18802

Supelco

2-Methyl-trans-aconitic acid sodium salt hydrate

≥95.0% (TLC)

Synonym(s):

(2E)-But-2-ene-1,2,3-tricarboxylic acid sodium salt hydrate, (E)-2-Butene-1,2,3-tricarbocylic acid sodium salt hydrate, (E)-2-Butene-1,2,3-tricarbocylic acid sodium salt hydrate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H8O6 · xNa+ · yH2O
Molecular Weight:
188.13 (anhydrous free acid basis)
Beilstein:
2416239
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.24

Quality Level

Assay

≥95.0% (TLC)

application(s)

clinical testing

format

neat

storage temp.

2-8°C

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Milko Velarde et al.
Journal of molecular biology, 391(3), 609-620 (2009-06-30)
3-Methylitaconate-Delta-isomerase (Mii) participates in the nicotinate fermentation pathway of the anaerobic soil bacterium Eubacterium barkeri (order Clostridiales) by catalyzing the reversible conversion of (R)-3-methylitaconate (2-methylene-3-methylsuccinate) to 2,3-dimethylmaleate. The enzyme is also able to catalyze the isomerization of itaconate (methylenesuccinate) to
A R Horswill et al.
Biochemistry, 40(15), 4703-4713 (2001-04-11)
Salmonella enterica serovar Typhimurium LT2 catabolizes propionate through the 2-methylcitric acid cycle, but the identity of the enzymes catalyzing the conversion of 2-methylcitrate into 2-methylisocitrate is unclear. This work shows that the prpD gene of the prpBCDE operon of this
J V Schloss et al.
Biochemistry, 23(20), 4572-4580 (1984-09-25)
alpha-Methyl-cis-aconitate (cis-2-butene-1,2,3-tricarboxylate) was converted only to alpha-methylisocitrate (3-hydroxybutane-1,2,3-tricarboxylate) by aconitases from beef liver or S. lipolytica. While the kinetic parameters of beef liver (cytoplasmic) or heart (mitochondrial) aconitases did not vary over the pH range 4.9-9 with the natural substrates
Tong Zhang et al.
Analytical chemistry, 84(4), 1994-2001 (2012-03-14)
In this study, we assessed three liquid chromatographic platforms: reversed phase (RP), aqueous normal phase (ANP), and hydrophilic interaction (HILIC) for the analysis of polar metabolite standard mixtures and for their coverage of urinary metabolites. The two zwitterionic HILIC columns
Tao Wu et al.
Journal of proteome research, 14(1), 447-456 (2014-09-24)
A number of metabolic conditions, including hypoglycemia, high blood pressure (HBP), dyslipidemia, nerve damage and amputation, and vision problems, occur as a result of uncontrolled blood glucose levels over a prolonged period of time. The different components of diabetic complications

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service