Skip to Content
Merck
All Photos(1)

Key Documents

18702

Sigma-Aldrich

Hydrogen bromide

puriss., ≥99.8%

Synonym(s):

Hydrobromic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HBr
CAS Number:
Molecular Weight:
80.91
Beilstein:
3587158
EC Number:
MDL number:
UNSPSC Code:
12142100

vapor density

2.71 (20 °C, vs air)

vapor pressure

334.7 psi ( 21 °C)

grade

puriss.

Assay

≥99.8%

impurities

≤0.1 ppm H2SO4
≤0.2% HCl
≤2 ppm Fe
≤5 ppm Br

bp

−67 °C (lit.)

mp

−87 °C (lit.)

SMILES string

Br

InChI

1S/BrH/h1H

InChI key

CPELXLSAUQHCOX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Packaging

Cylinder with net ~450 g

Other Notes

Sales restrictions may apply

replaced by

Product No.
Description
Pricing

Signal Word

Danger

Hazard Statements

Hazard Classifications

Press. Gas Compr. Gas - Skin Corr. 1A - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

2A - Gases

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service