Skip to Content
Merck
All Photos(1)

Key Documents

17786

Sigma-Aldrich

Geraldol

≥97.0% (HPLC)

Synonym(s):

3′-Methoxy-3,7,4′-trihydroxyflavone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H12O6
CAS Number:
Molecular Weight:
300.26
Beilstein:
1657077
EC Number:
MDL number:
UNSPSC Code:
12352200

Assay

≥97.0% (HPLC)

SMILES string

COc1cc(ccc1O)C2=C(O)C(=O)c3ccc(O)cc3O2

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service