Skip to Content
Merck
All Photos(1)

Key Documents

11105

Sigma-Aldrich

Artemisia ketone

purum, ≥97.0% (GC)

Synonym(s):

3,3,6-Trimethyl-1,5-heptadien-4-one

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H16O
CAS Number:
Molecular Weight:
152.23
Beilstein:
1701720
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

grade

purum

Assay

≥97.0% (GC)

refractive index

n20/D 1.466

density

0.869 g/mL at 20 °C (lit.)

SMILES string

C\C(C)=C\C(=O)C(C)(C)C=C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service