Skip to Content
Merck
All Photos(1)

Documents

07553

Supelco

Diethyl maleate

analytical standard

Synonym(s):

Maleic acid diethyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C2H5OCOCH=CHCOOC2H5
CAS Number:
Molecular Weight:
172.18
Beilstein:
1100825
EC Number:
MDL number:
UNSPSC Code:
12352100
NACRES:
NA.24

grade

analytical standard

Quality Level

vapor density

5.93 (vs air)

vapor pressure

1 mmHg ( 14 °C)

Assay

≥96.5% (GC)

autoignition temp.

662 °F

shelf life

limited shelf life, expiry date on the label

refractive index

n20/D 1.440-1.442
n20/D 1.441 (lit.)

bp

225 °C (lit.)

mp

−10 °C (lit.)

density

1.064 g/mL at 25 °C (lit.)

application(s)

cleaning products
cosmetics
food and beverages
personal care

format

neat

SMILES string

CCOC(=O)\C=C/C(=O)OCC

InChI

1S/C8H12O4/c1-3-11-7(9)5-6-8(10)12-4-2/h5-6H,3-4H2,1-2H3/b6-5-

InChI key

IEPRKVQEAMIZSS-WAYWQWQTSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Eye Irrit. 2 - Skin Sens. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

200.1 °F - closed cup

Flash Point(C)

93.4 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service