05400
Ala-Trp
≥99.0% (TLC)
Synonym(s):
L-Alanyl-L-tryptophan
About This Item
Quality Level
Assay
≥99.0% (TLC)
form
powder
optical activity
[α]20/D +15.5±0.5°, c = 2.7% in H2O
reaction suitability
reaction type: solution phase peptide synthesis
technique(s)
HPLC: suitable
color
colorless to white
application(s)
peptide synthesis
SMILES string
C[C@H](N)C(=O)N[C@@H](Cc1c[nH]c2ccccc12)C(O)=O
InChI
1S/C14H17N3O3/c1-8(15)13(18)17-12(14(19)20)6-9-7-16-11-5-3-2-4-10(9)11/h2-5,7-8,12,16H,6,15H2,1H3,(H,17,18)(H,19,20)/t8-,12-/m0/s1
InChI key
WUGMRIBZSVSJNP-UFBFGSQYSA-N
Looking for similar products? Visit Product Comparison Guide
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service