Skip to Content
Merck
All Photos(1)

Key Documents

03756

Sigma-Aldrich

(S)-2-Boc-1,2,3,4-tetrahydronorharmane-3-carboxylic acid

≥96.0% (HPLC)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H20N2O4
CAS Number:
Molecular Weight:
316.35
Beilstein:
691288
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

Pricing and availability is not currently available.

Assay

≥96.0% (HPLC)

SMILES string

CC(C)(C)OC(=O)N1Cc2[nH]c3ccccc3c2C[C@H]1C(O)=O

InChI

1S/C17H20N2O4/c1-17(2,3)23-16(22)19-9-13-11(8-14(19)15(20)21)10-6-4-5-7-12(10)18-13/h4-7,14,18H,8-9H2,1-3H3,(H,20,21)/t14-/m0/s1

InChI key

FHEPEWKHTOVVAT-AWEZNQCLSA-N

Looking for similar products? Visit Product Comparison Guide


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service