Skip to Content
Merck
All Photos(1)

Documents

01870

Supelco

Silica gel 60 ADAMANT on TLC plates

with fluorescence indicator 254 nm

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
41115711
PubChem Substance ID:
NACRES:
NC.07

Quality Level

feature

fluorescent indicator

technique(s)

thin layer chromatography (TLC): suitable

layer thickness

250 μm ± 30 μm

plate size

20 cm × 20 cm

particle size

7.0-11.0 μm

pore size

60 Å medium pore diameter

SMILES string

O=[Si]=O

InChI

1S/O2Si/c1-3-2

InChI key

VYPSYNLAJGMNEJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Adamant is a trademark of Macherey-Nagel GmbH & Co., KG

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service