Skip to Content
Merck
All Photos(1)

Documents

PBMK06210

Millipore

PES Membrane Filter, 300 kDa NMWCO

Biomax®, filter diam. 63.5 mm

Synonym(s):

Biomax PES ultrafiltration membrane discs, Ultrafiltration Discs, 300 kDa NMW

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41104903
eCl@ss:
32031602
NACRES:
NB.24

material

polyethersulfone membrane

Quality Level

manufacturer/tradename

Biomax®

technique(s)

protein clean-up: suitable (desalting)
protein purification: suitable (concentration)

filter diam.

63.5 mm

filtration area

28.7 cm2

matrix

Biomax®

pore size

300 kDa NMWCO

shipped in

ambient

General description

Ultrafiltration membranes are generally composed of regenerated cellulose or polyethersulfone (PES). The sample compatibility and the molecular weight (MW) of the macrosolute that is to be retained determine the choice of membrane and its nominal molecular weight cut-off (NMWCO), respectively. As a rule, the NMWCO should be 4-5 times smaller than the molecular weight of the solute to be retained when using Biomax® PES membranes.

Application

Ultrafiltration Discs, 300 kDa NMW has been used for Kaposi′s sarcoma-associated herpesvirus (KSHV) purification.

Legal Information

BIOMAX is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jian-Jun Wu et al.
Journal of virology, 90(4), 1741-1756 (2015-12-08)
We recently showed that the interaction between Kaposi's sarcoma-associated herpesvirus (KSHV) tegument proteins ORF33 and ORF45 is crucial for progeny virion production, but the exact functions of KSHV ORF33 during lytic replication were unknown (J. Gillen, W. Li, Q. Liang

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service