Skip to Content
Merck
All Photos(4)

Documents

MATFLAGELLIN

Millipore

NEP Control Flagellin- PyroMAT® System MAT

Non-endotoxin pyrogen control of flagellin for use in monocyte activation test

Synonym(s):

NEP, NEP Control

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116130
eCl@ss:
32160606
NACRES:
NA.24

Assay

≥95% (SDS-PAGE)

Quality Level

usage

sufficient for 10 tests

feature

Non-endotoxin pyrogen control of flagellin for use in monocyte activation test

manufacturer/tradename

PyroMAT®

application(s)

pharmaceutical
pyrogen testing

compatibility

configured for PyroDetect System
configured for PyroMAT® System

shipped in

cooler

storage temp.

−20°C

suitability

pyrogenic bacteria

Related Categories

General description

The PyroMAT® System uses cryopreserved Mono-Mac-6 (MM6) human monocytic cells as a source of monocytes. The response to a full range of pyrogens, including endotoxins and non-endotoxin pyrogens (NEPs)is determined by the measurement of interleukin-6 (IL-6) produced by the Mono-Mac-6 cells using ELISA.
NEP control flagellin is a preparation of flagellin from Salmonella typhimurium and supplied in a lyophilized form. Flagellin is a non-endotoxin pyrogen (NEP) that activates the TLR5 receptor of monocytes and can be used as a positive control with the PyroMAT® System (Monocyte Activation Test for pyrogen detection).

Specificity

TLR-5 agonist

Application

NEP control of flagellin can be used as a positive control with the PyroMAT® System. It is to be used for Product Specific Validation (PSV) and routine testing to show that the system can detect non-endotoxin pyrogen contaminants in pharmaceutical products.

Features and Benefits

Endotoxin-free product at concentration of use in MAT Purity: ≥ 95% (SDS-PAGE)

Storage and Stability

Store unopened vials at -20°C
Do not use unopened vials after expiration date
Store resuspended aliquots at -20°C
Shelf life of aliquots can be found on the technical datasheet

Legal Information

PYROMAT is a registered trademark of Merck KGaA, Darmstadt, Germany

also commonly purchased with this product

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service