Skip to Content
Merck
All Photos(1)

Key Documents

MABS1161

Sigma-Aldrich

Anti-PYCR1 Antibody, clone 2D9.1

clone 2D9.1, from mouse

Synonym(s):

Pyrroline-5-carboxylate reductase 1, mitochondrial, PYCR1, P5C reductase 1, P5CR 1

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€290.00

€290.00


Estimated to ship on11 April 2025Details


Request a Bulk Order

Select a Size

Change View
100 μG
€290.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€290.00


Estimated to ship on11 April 2025Details


Request a Bulk Order

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

2D9.1, monoclonal

species reactivity

human

technique(s)

immunohistochemistry: suitable (paraffin)
western blot: suitable

isotype

IgG2bκ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... PYCR1(5831)

General description

Pyrroline-5-carboxylate reductase 1, mitochondrial (EC 1.5.1.2; UniProt P32322; also known as P5C reductase 1, P5CR 1) is encoded by the PYCR1 (also known as ARCL2B, ARCL3B) gene (Gene ID 5831) in human. Pyrroline-5-carboxylate (P5C) reductase (PYCR1) is a mitochondrial enzyme that utilizes NAD(P)H as cofactor and catalyzes the convertion of P5C to L-proline, as well as the coversion of Δ1-piperideine-6-carboxylate (P6C) to L-pipecolic acid, both of which are intermediates in the catabolism pathway of the essential amino acid L-lysine. PYCR1 gene mutations are associated with the autosomal recessive disorder cutis laxa type II, menifested with premature aging features such as joint laxity and wrinkled/lax skin, as well as developmental delay.

Specificity

Expected to react with spliced isoforms 1 and 3, but not isoform 2, reported by UniProt P32322).

Immunogen

GST-tagged recombinant protein corresponding to the C-terminal of human PYCR1.

Application

Anti-PYCR1 Antibody, clone 2D9.1 is an antibody against PYCR1 for use in Western Blotting, Immunohistochemistry (Paraffin).

Quality

Evaluated by Western Blotting in K562 cell lysate.

Western Blotting Analysis: 0.5 µg/mL of this antibody detected PYCR1 in 10 µg of K562 cell lysate.

Target description

~33 kDa observed. 33.36 kDa (isoform 1) and 35.98 kDa (isoform 2) calculated. Uncharacterized band(s) may appear in some lysates. Uncharacterized band(s) may appear in some lysates.

Physical form

Format: Purified

Other Notes

Concentration: Please refer to lot specific datasheet.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service