Skip to Content
Merck
All Photos(4)

Key Documents

MABS1156

Sigma-Aldrich

Anti-Fibrocystin Antibody, clone 18

clone 18, from mouse

Synonym(s):

Fibrocystin, Polycystic kidney and hepatic disease 1 protein, Polyductin, Tigmin, Fibrocystin

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€445.00

€445.00


Estimated to ship on03 March 2025Details


Request a Bulk Order

Select a Size

Change View
100 μG
€445.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€445.00


Estimated to ship on03 March 2025Details


Request a Bulk Order

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

18, monoclonal

species reactivity

human, rat

technique(s)

immunocytochemistry: suitable
immunohistochemistry: suitable (paraffin)

isotype

IgG2aκ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... PKHD1(5314)

General description

Fibrocystin (UniProt P08F94; also known as Polycystic kidney and hepatic disease 1 protein, Polyductin, Tigmin, TIG multiple domains 1, Tigmin) is encoded by the PKHD1 (also known as ARPKD, FCYT, TIGM1) gene (Gene ID 5314) in human. Autosomal recessive polycystic kidney disease (ARPKD) is the most common cause of hereditary childhood PKD. ARPKD is caused by mutations in the gene PKHD1, which encodes fibrocystin/polyductin (FCP), while mutations in two other genes PKD1 and PKD2 that code polycystin-1 (PC1) and polycystin-2 (PC2), respectively, are the cause of autosomal dominant PKD (ADPKD). These three PKD proteins have been localized to renal primary cilia, where the PC1/PC2 complex acts as a flow sensor on the cilium. Fibrocystin is known to complex with PC2, but its biological role is unclear, While Uniprot (P08F94) documents two alternatively spliced human fibrocytin isoforms, multiple lines of evidence indicate that many more spliced mRNA transcripts exist and that aberrant mRNA splicing may represent a pathogenic mechanism in ARPKD in addition to gene mutations.

Specificity

Expected to react with both spliced isoforms.

Immunogen

Epitope: N-terminal half
Recombinant protein corresponding to human Fibrocystin fragment from the N-terminal half.

Application

Immunohistochemistry Analysis: A 1:50 dilution from a representative lot detected Fibrocystin in human kidney cortex, rat kidney cortex, and rat kidney medulla tissue.
Immunocytochemistry Analysis: A representative lot detected fibrocystin immunoreactivity associated with primary cilia of cultured normal human kidney epithelial cells (Courtesy of Dr Christopher Ward, University of Kansas Medical Center).
Research Category
Signaling
Research Sub Category
Signaling Neuroscience
This Anti-Fibrocystin Antibody, clone 18 is validated for use in Immunohistochemistry (Paraffin), Immunocytochemistry for the detection of Fibrocystin.

Quality

Evaluated by Immunohistochemistry in human kidney medulla tissue.

Immunohistochemistry Analysis: A 1:50 dilution of this antibody detected Fibrocystin in human kidney medulla tissue.

Target description

~371 to 446 kDa calculated

Physical form

Format: Purified
Protein G Purified
Purified mouse monoclonal IgG2aκ antibody in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service