MABN874
Anti-KLK6 Antibody, clone 27-4
clone 27-4, from mouse
Synonym(s):
Kallikrein-6, hK6, Neurosin, Protease M, Serine protease 18, Serine protease 9, SP59, Zyme
About This Item
biological source
mouse
antibody form
purified immunoglobulin
antibody product type
primary antibodies
clone
27-4, monoclonal
species reactivity
human
technique(s)
ELISA: suitable
affinity chromatography: suitable
western blot: suitable
isotype
IgG2bκ
NCBI accession no.
UniProt accession no.
target post-translational modification
unmodified
Gene Information
human ... KLK6(5653)
General description
Specificity
Immunogen
Application
ELISA Analysis: Representative lots were employed as the capture antibody in combination with biotin-conjugated clone E24 (Cat. No. MABN873) as the detection antibody for analyzing kallikrein-6 levels in ovarian cytosolic extracts as well as ascites fluids from ovarian cancer patients (Shan, S.J., et al. (2007). Br. J. Cancer. 96(2):362-372; Luo, L.Y., et al. (2006). Tumor Biol. 27:227-234).
Immunoaffinity Purification: A representative lot was conjugated to resins and employed for the affinity purification of kallikrein-6 from human cerebrospinal fluids (CSFs) and ascites fluids (Kuzmanov, U., et al. (2009). Mol. Cell. Proteomics. 8(4):791-798).
Quality
Western Blotting Analysis: 0.5 µg/mL of this antibody detected kallikrein-6 in 10 µg of human ovary tissue lysate.
Target description
Physical form
Other Notes
Not finding the right product?
Try our Product Selector Tool.
Storage Class Code
12 - Non Combustible Liquids
WGK
nwg
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service