MABN448
Anti-ENC-1 Antibody, clone 1H8.1
clone 1H8.1, from mouse
Synonym(s):
Ectoderm-neural cortex protein 1, ENC-1, Kelch-like protein 37, Nuclear matrix protein NRP/B, p53-induced gene 10 protein
About This Item
biological source
mouse
Quality Level
antibody form
purified antibody
antibody product type
primary antibodies
clone
1H8.1, monoclonal
species reactivity
human, rat
technique(s)
immunohistochemistry: suitable
western blot: suitable
isotype
IgG2aκ
NCBI accession no.
UniProt accession no.
shipped in
wet ice
target post-translational modification
unmodified
Gene Information
human ... ENC1(8507)
General description
Immunogen
Application
Immunohistochemistry Analysis: A 1:50 dilution from a representative lot detected ENC-1 in human cerebellum and cerebral cortex tissue.
Quality
Western Blotting Analysis: 1 µg/mL of this antibody detected ENC-1 in 10 µg of human hippocampus tissue.
Target description
Linkage
Physical form
Other Notes
Not finding the right product?
Try our Product Selector Tool.
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service