Skip to Content
Merck
All Photos(2)

Key Documents

MABN378

Sigma-Aldrich

Anti-Potassium Channel Kv10.1 Antibody, clone 56

clone 56, from mouse

Synonym(s):

Potassium voltage-gated channel subfamily H member 1, Ether-a-go-go potassium channel 1, EAG channel 1, h-eag, hEAG1, Voltage-gated potassium channel subunit Kv10.1, hEag1 Potassium channel, eag1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

56, monoclonal

species reactivity

rat, human

technique(s)

activity assay: suitable
immunofluorescence: suitable
immunohistochemistry: suitable

isotype

IgG2bκ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... KCNH1(3756)

General description

Potassium Channel Kv10.1 is also known as Potassium voltage-gated channel subfamily H member 1, Ether-a-go-go potassium channel 1 (EAG channel 1, h-eag, or hEAG1) and Voltage-gated potassium channel subunit Kv10.1. Potassium Channel Kv10.1 is a pore-forming (alpha) subunit of voltage-gated, non-inactivating, delayed rectifier potassium channel. Potassium Channel Kv10.1 mediates IK(NI) current in myoblasts, and can be modulated by cAMP as well as subunit assembly. Potassium Channel Kv10.1 is highly expressed in myoblasts at the onset of fusion and brain tissue. It is not found in normal epithelial cells, but has been detected in HeLa (cervical carcinoma), SH-SY5Y (neuroblastoma) and MCF-7 (epithelial tumor) cells.

Specificity

This antibody recognizes the extracellular, C-terminus domain of Potassium Channel Kv10.1. Additionally, this antibody does not cross react with hEag2 (Gomez-Varela, D., et al. (2007). Cancer Res. 67(15):7343-7349.).

Immunogen

Epitope: Extracellular domain
Recombinant protein corresponding to the extracellular domain of human Potassium Channel Kv10.1.

Application

Detect Kv10.1 using this mouse monoclonal antibody, Anti-Potassium Channel Kv10.1 Antibody, clone 56 validated for use in IHC, Activity Assay & Immunofluorescence.
Immunohistochemistry Analysis: A 1:250 dilution from a representative lot detected Potassium Channel Kv10.1 in human thalamus and cerebellum tissues.

Immunocytochemistry Analysis: A representative lot detected Potassium Channel Kv10.1 in transfected CHO-K1 and HEK293 cells (Gomez-Varela, D., et al. (2007). Cancer Res. 67(15):7343-7349.).

Activity Assay Analsyis: A representative lot from an independent laboratory inhibited anchorage-independent cancer cell growth in vitro (Gomez-Varela, D., et al. (2007). Cancer Res. 67(15):7343-7349.; Chen, Y., et al. (2011). PLoS One. 6(5):e19257.).

Activity Assay Analysis: A representative lot from independent laboratories inhibited Potassium Channel Kv10.1 currents in transfected HEK293 cells (Gomez-Varela, D., et al. (2007). Cancer Res. 67(15):7343-7349.).

Activity Assay Analysis: A representative lot from an independent laboratory inhibited cellular proliferation and
migration of certain Acute myeloid leukemia cell lines and primary cultured AML cells in vitro (Agarwal, J. R., et al. (2010). Mol Cancer. 9:18.).
Research Category
Neuroscience
Research Sub Category
Signaling Neuroscience

Quality

Evaluated by Immunohistochemistry Analysis in rat frontal lobe tissue.

Immunohistochemistry Analysis: A 1:50 dilution of this antibody detected Potassium Channel Kv10.1 in rat frontal lobe tissue.

Target description

111 kDa calculated

Physical form

Format: Purified
Protein G Purified
Purified mouse monoclonal IgG2bκ in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
Rat brain tissue

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service