Skip to Content
Merck
All Photos(2)

Key Documents

MABF240

Sigma-Aldrich

Anti-hGITR Antibody, clone 108-17

clone 108-17, from mouse

Synonym(s):

Tumor necrosis factor receptor superfamily member 18, Activation-inducible TNFR family receptor, Glucocorticoid-induced TNFR-related protein, CD357

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€248.00

€248.00


Estimated to ship on16 April 2025Details


Request a Bulk Order

Select a Size

Change View
100 μG
€248.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€248.00


Estimated to ship on16 April 2025Details


Request a Bulk Order

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

108-17, monoclonal

species reactivity

mouse, human, rat

technique(s)

western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... TNFRSF18(8784)

General description

Glucocorticoid-induced TNFR-related protein (hGITR) is also called Tumor necrosis factor receptor superfamily member 18 (TNFRSF18), Activation-inducible TNFR family receptor (AITR) and CD357. hGITR is a receptor for TNFSF18 and is activated by NF-kappa-B through the TRAF2/NIK pathway. hGITR is thought to be involved in activated T-lymphocyte and endothelial cell interactions, as well as regulation of T-cell receptor-mediated cell death. hGITR is expressed in lymph node, peripheral blood leukocytes and at low levels in the spleen.

Immunogen

Recombinant protein corresponding to human hGITR.

Application

Anti-hGITR Antibody, clone 108-17 is a highly specific mouse monoclonal antibody & that targets GITR & has been tested in western blotting.
Research Category
Inflammation & Immunology
Research Sub Category
Immunoglobulins & Immunology
Western Blotting Analysis: 1.0 µg/mL from a representative lot detected hGITR in 10 µg of mouse lymph node, rat spleen, and rat thymus tissue lysate.

Quality

Evaluated by Western Blotting in human spleen tissue lysate.

Western Blotting Analysis: 1.0 µg/mL of this antibody detected hGITR in 10 µg of human spleen tissue lysate.

Target description

~ 26 kDa observed. Multiple bands can be observed due to GITR being a monomer & homeodimer in some lysates. Uncharacterized band(s) may appear in some lysates.

Physical form

Format: Purified
Protein A purified
Purified mouse monoclonal IgG1κ in buffer containing PBS with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service