Skip to Content
Merck
All Photos(1)

Key Documents

MABE1167

Sigma-Aldrich

Anti-SPO11 Antibody, clone 180

clone 180, from mouse

Synonym(s):

Meiotic recombination protein SPO11

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€392.00

€392.00


Estimated to ship on28 April 2025Details


Request a Bulk Order

Select a Size

Change View
100 μG
€392.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702

€392.00


Estimated to ship on28 April 2025Details


Request a Bulk Order

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

180, monoclonal

species reactivity

mouse

technique(s)

immunocytochemistry: suitable
immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG2aκ

NCBI accession no.

UniProt accession no.

shipped in

ambient

target post-translational modification

unmodified

Gene Information

mouse ... Spo11(26972)

General description

Meiotic recombination protein SPO11 (UniProt Q9WTK8) is encoded by the Spo11 gene (Gene ID 26972) in murine species. SPO11 is a widely conserved protein that is involved in meiotic recombination and mediates DNA cleavage that forms the double-strand breaks that initiate meiotic recombination. Active site of SPO11 contains a tyrosine, which ligates and dissociates with DNA to promote break formation. One SPO11 protein is involved per strand of DNA, hence, two SPO11 proteins are involved in each double stranded break event. High levels of SPO11 are found only in the testis where expression is restricted primarily to meiotic germ cells and it is not expressed in spermatogonia. Very low levels of SPO11 have been reported in thymus, brain, and oocytes of embryonic ovary, but it is not detected in adult ovary. Mouse and human SPO11 exhibit about 82% sequence homology. Ref.: Keeney, S., et al. (1997). Cell 375-384.

Specificity

Clone 180 detected SPO11alpha and SPO11beta in wild-type, but not Spo11-knockout mouse testes lysates by IP-Western blotting.

Immunogen

Recombinant mouse SPO11 with N-terminal S-tag and C-terminal His-tag.

Application

Anti-SPO11, clone 180, Cat. No. MABE1167, is a highly specific mouse monoclonal antibody that targets SPO11 and has been tested in and Immunocytochemistry, Immunoprecipitation, and Western Blotting.
Immunoprecipitation Analysis: 3 µg from a representative lot immunoprecipitated SPO11alpha and SPO11beta from wild-type, but not Spo11-knockout mouse testes lysates (Courtesy of the Keeney Lab, MSKCC, New York, U.S.A.).

Western Blotting Analysis: A representative lot detected SPO11alpha and SPO11beta in immunoprecipitate from wild-type, but not SPO-knockout, mouse testes using the same antibody (Courtesy of the Keeney Lab, MSKCC, New York, U.S.A.).

Immunocytochemistry Analysis: A representative lot immunostained SPO11 in meiotic prophase I leptotene cells by fluorescence immunocytochemistry staining of chromosome spreads from isolated mouse spermatocytes hypotonically swollen, fixed with 1% formaldehyde and permeabilized with 0.1% Triton X-100 (Alpatov, R., et al. (2014). Cell. 157(4):869-881).

Immunoprecipitation Analysis: A representative lot immunoprecipitated SPO11alpha and SPO11beta as well as SPO11-oligo complexes from mouse testes lysates. Much elevated and greatly reduced SPO11-oligo complexes were found in Atm-/- and Dmc-/- testes IP, respectively, when comparing to wild-type testes IP (Lange, J., et al. (2011). Nature. 479(7372):237-240).

Western Blotting Analysis: Representative lots detected SPO11alpha and SPO11beta in immunoprecipitates obtained with the same antibody from mouse testes. Much reduced SPO11 was found in Atm-/- and Dmc-/- testes IPs due to meiotic arrest at prophase I (Biswas, U., et al. (2013). PLoS Genet. 9(12):e1003985; Lange, J., et al. (2011). Nature. 479(7372):237-240).
Research Category
Epigenetics & Nuclear Function

Quality

Identity Confirmation by Isotyping Test.

Isotyping Analysis: The identity of this monoclonal antibody is confirmed by isotyping test to be mouse IgG2aκ.

Target description

~44/40 kDa (SPO11beta/SPO11alpha) observed. Uncharacterized bands may be observed in some lysate(s).

Physical form

Format: Purified
Protein G purified.
Purified mouse IgG2aκ in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tina N Tran et al.
Biology of reproduction, 101(2), 347-359 (2019-05-11)
A major challenge in medical genetics is to characterize variants of unknown significance (VUS). Doing so would help delineate underlying causes of disease and the design of customized treatments. Infertility has presented an especially difficult challenge with respect to not

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service