Skip to Content
Merck
All Photos(1)

Key Documents

MABE1052

Sigma-Aldrich

Anti-GMPS Antibody, clone 1G6.1

clone 1G6.1, from mouse

Synonym(s):

GMP synthase [glutamine-hydrolyzing], Glutamine amidotransferase, GMP synthetase

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
€238.00

€238.00


Estimated to ship on09 April 2025Details


Request a Bulk Order

Select a Size

Change View
100 μL
€238.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702

€238.00


Estimated to ship on09 April 2025Details


Request a Bulk Order

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

1G6.1, monoclonal

species reactivity

human

technique(s)

western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

ambient

target post-translational modification

unmodified

Gene Information

human ... GMPS(8833)

General description

GMP synthase [glutamine-hydrolyzing] (EC 6.3.5.2; UniProt P49915; also known as Glutamine amidotransferase, GMP synthetase) is encoded by the GMPS gene (Gene ID 8833) in human. De novo biosynthesis of Guanosine 5′-monophosphate (GMP) is accomplished by reactions catalyzed by several enzymes. Inositol monophosphate (IMP) dehydrogenase 1 and 2 (IMPDH1 and IMPDH2) convert IMP into xanthosine monophosphate (XMP), GMP synthetase (GMPS) then converts XMP into GMP, while guanosine monophosphate reductase (GMPR) catalyzes the reversible conversion of GMP to IMP to fuel back into both the AMP and GMP synthesis pathways. GMPS is a bifunctional two-domain N-type pyrophosphatase whose N-terminal glutaminase domain (a.a. 27-216) generates ammonia from glutamine, while its C-terminal synthetase domain (a.a. 27-435) adenylylates XMP on the xanthine C2 oxygen, thereby priming XMP for attack by a nitrogen nucleophile to form GMP. Human GMPS activity is subjected to an allosteric regulation and displays positive cooperativity for the substrate XMP. GMPS is normally sequestered in the cytoplasm, but translocates to the nucleus in response to genotoxic stress or nucleotide deprivation. GMPS nuclear translocation plays an important role in USP7-mediated stabilization of p53, where p53 is transferred from MDM2 to the GMPS-USP7 deubiquitylation complex.

Specificity

Clone 1G6.1 targets an epitope within the GMPS ATP-PPase domain region present in both spliced isoforms of human GMPS reported by UniProt (P49915).

Immunogen

GST-tagged recombinant human GMPS ATP-PPase domain fragment.

Application

Anti-GMPS, clone 1G6.1, Cat. No. MABE1052, is a highly specific mouse monoclonal antibody that targets GMPS and has been tested in Western Blotting.
Research Category
Epigenetics & Nuclear Function

Quality

Evaluated by Western Blotting in HeLa cell lysate.

Western Blotting Analysis: A 1:2,000 dilution of this antibody detected GMPS in 10 µg of HeLa cell lysate.

Target description

~70 kDa observed. 76.58/65.80 kDa (isoform 1/2) calculated (Met1 removed). Uncharacterized bands may be observed in some lysate(s).

Physical form

Format: Purified
Protein G purified
Purified mouse IgG1 in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service