Skip to Content
Merck
All Photos(1)

Key Documents

MABC531

Sigma-Aldrich

Anti-Somatotropin/GH1 Antibody, clone 1H1.1

clone 1H1.1, from mouse

Synonym(s):

Somatotropin, Growth hormone, GH, GH-N, Growth hormone 1, Pituitary growth hormone, -Somatotropin/GH1

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€429.00

€429.00


Estimated to ship on24 April 2025Details


Request a Bulk Order

Select a Size

Change View
100 μG
€429.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.43

€429.00


Estimated to ship on24 April 2025Details


Request a Bulk Order

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

1H1.1, monoclonal

species reactivity

human

technique(s)

western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... GH1(2688)

General description

Somatotropin (UniProt P01241; also known as GH, GH-N, Growth hormone 1, hGH-N, Pituitary growth hormone) is encoded by the GH1 (also known as GH, GHN, IGHD1B) gene (Gene ID 8856) in human. Somatotropin is a peptide hormone that plays an important role in growth control by inducing IGF-1 secretion in liver and other tissues. It stimulates myoblast differentiation and proliferation, as well as amino acids uptake and protein synthesis in muscle and other tissues. Acromegaly is a rare endocrine syndrome caused by a GH-secreting pituitary adenoma. Acromegaly may also be rarely associated with a hypothalamic or an ectopic GH-releasing hormone-producing tumor.

Specificity

Expected to react with isoforms 1, 3, and 4. Reactivity to isoforms 2 and 5 have not been tested.

Immunogen

GST-tagged recombinant protein corresponding to human Somatotropin/GH1.

Application

Anti-Somatotropin/GH1 Antibody, clone 1H1.1 is an antibody against Somatotropin/GH1 for use in Western Blotting.
Research Category
Apoptosis & Cancer
Research Sub Category
Apoptosis - Additional

Quality

Evaluated by Western Blotting in human pituitary tissue lysate.

Western Blotting Analysis: 0.5 µg/mL of this antibody detected Somatotropin/GH1 in 10 µg of human pituitary tissue lysate.

Target description

~25 kDa observed. This protein has several isoforms from ~20-25 kDa

Physical form

Format: Purified
Protein G Purified
Purified mouse monoclonal IgG1κ antibody in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service