Skip to Content
Merck
All Photos(2)

Documents

MAB374-AF488

Sigma-Aldrich

Anti-GAPDH Antibody, clone 6C5, Alexa Fluor 488 conjugate

clone 6C5, from mouse, ALEXA FLUOR 488

Synonym(s):

Glyceraldehyde-3-phosphate dehydrogenase, GAPDH, Peptidyl-cysteine S-nitrosylase GAPDH, GAPDH

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

conjugate

ALEXA FLUOR 488

antibody form

purified antibody

antibody product type

primary antibodies

clone

6C5, monoclonal

species reactivity

mouse, human

species reactivity (predicted by homology)

cat (based on 100% sequence homology), fish (based on 100% sequence homology), canine (based on 100% sequence homology), porcine (based on 100% sequence homology), rat (based on 100% sequence homology), rabbit (based on 100% sequence homology)

technique(s)

immunocytochemistry: suitable

isotype

IgG1

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... GAPDH(2597)
mouse ... Gapdh(14433)
rabbit ... Gapdh(100009074)

General description

Glyceraldehyde 3-phosphate dehydrogenase (GAPDH) is an ubiquitous glycolytic enzyme present in reasonably high levels in almost all tissues. The complete and functional enzyme is a tetramer with each of four identical subunits occupying the vertex of a tetrahedron. Its binding domains also include one principally interacting with NAD+ and another interacting with glyceraldehyde 3-phosphate (GAP). As a ′house-keeping′ enzyme, it catalyzes the synthesis of 1,3-biphosphoglycerate, a "high energy" intermediate used for the synthesis of ATP. Besides its cytoplasmic role in metabolism, it is also involved in the initial stages of apoptosis or oxidative stress response where it is translocated to the nucleus. Such actions may reflect the role of GAPDH in DNA repair or as one nuclear carrier for apoptotic molecules. GAPDH can also bind specifically to proteins implicated in the pathogenesis of a variety of neurodegenerative disorders, including the beta-amyloid precursor protein and the Huntingtin protein where reduced function of GAPDH is associated with Alzheimer′s and Huntington′s disease fibroblasts. GAPDH has also been identified as a potential target for nitric oxide (NO)-mediated cellular toxicity.

Specificity

It has been reported that this antibody does not react with GAPDH from E. coli.

Application

This Anti-GAPDH Antibody, clone 6C5, Alexa Fluor 488 conjugate is validated for use in Immunocytochemistry for the detection of GAPDH.

Quality

Evaluated by Immunocytochemistry in A431 cells.

Immunocytochemistry Analysis: A 1:100 dilution of this antibody detected GAPDH in A431 cells.

Target description

~38 kDa observed

Physical form

Purified mouse conjugate antibody in buffer containing PBS 15 mg/ml BSA and 0.1% Sodium Azide.

Other Notes

Concentration: Please refer to lot specific datasheet.

Legal Information

ALEXA FLUOR is a trademark of Life Technologies

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Carolina Cano Macip et al.
Cellular reprogramming, 26(1), 24-32 (2024-02-21)
Aging is a complex progression of changes best characterized as the chronic dysregulation of cellular processes leading to deteriorated tissue and organ function. Although aging cannot currently be prevented, its impact on life- and healthspan in the elderly can potentially

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service