HG00C117
MYGP Agar
final pH 6.2±0.2 (25 °C, after autoclaving)
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41116105
Recommended Products
composition
Agar, 27.0 g/L
Dextrose, 11.0 g/L
Maltose, 3.0 g/L
Peptone, 5.0 g/L
Yeast Extract, 3.0 g/L
packaging
poly bottle of 12 × 500 g (HG00C117.500)
final pH
6.2±0.2 (25 °C, after autoclaving)
storage temp.
15-25°C
Application
A medium suitable for the cultivation, isolation and maintenance of yeast, moulds and other aciduric microorganisms.
Preparation Note
Suspend 51g in 1 litre demineralised water. Boil whilst stirring until completely dissolved. Autoclave at 121°C for 15 minutes. Cool to 45 - 50°C, mix well and pour plates.
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 2
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service