Skip to Content
Merck
All Photos(1)

Key Documents

FR180380143

Sigma-Aldrich

K1-030 Black COOH PS 10%

Sign Into View Organizational & Contract Pricing

Select a Size

80380143-0010
€387.00

€387.00


Estimated to ship on11 April 2025Details

For orders above 5 units please use the Request a Bulk Order button.


Select a Size

Change View
80380143-0010
€387.00

About This Item

UNSPSC Code:
13102026

€387.00


Estimated to ship on11 April 2025Details

For orders above 5 units please use the Request a Bulk Order button.

Quality Level

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service