Skip to Content
Merck
All Photos(3)

Documents

EZFITMIC01

Millipore

EZ-Fit® Manifold

1-place, For Microfil® funnel and membranes

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116129
NACRES:
NB.24

material

stainless steel

Sterilization Compatibility

autoclavable compatible

packaging

pkg of 1 unit

manufacturer/tradename

EZ-Fit®

input

sample type pharmaceutical(s)
sample type cosmetics
sample type beer
sample type bottled water

application(s)

bioburden testing
cosmetics
environmental
food and beverages
water monitoring

shipped in

ambient

Related Categories

General description

EZ-Fit® Manifold helps to optimize the workflow and take control of the contamination risks. Every single component of the manifold can be removed for easy cleaning and autoclaving, thus preventing the formation of biofilm. The 2-way valve design prevents the user from accidentally touching the filtration head while opening or closing the valve. A low height design increases comfort when used in laminar flow hoods. The new Microfil® head design improves ease of use, reduces maintenance, and limits the risk of contamination. A continuous rim allows easy and safe membrane transfer with forceps, reducing the risk of touching the membrane filtration area. The porous filtration support can be removed simply by pressing down anywhere around its edge.

Application

The EZ-Fit® filtration Manifold is used for universal laboratory filtrations and supports the filtration of one test sample. The manifold is suitable to be used with Microfil® filtration units.

Legal Information

EZ-FIT is a registered trademark of Merck KGaA, Darmstadt, Germany
MICROFIL is a registered trademark of Merck KGaA, Darmstadt, Germany

configured for

Product No.
Description
Pricing

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Related Content

Microfil® filtration system ensures microbiological quality assurance in routine testing of raw materials.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service