Skip to Content
Merck
All Photos(1)

Key Documents

AP503R

Sigma-Aldrich

Goat Anti-Mouse IgG γ chain Antibody, TRITC conjugate, Species Adsorbed

Chemicon®, from goat

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
€208.00

€208.00


Estimated to ship on30 April 2025Details


Request a Bulk Order

Select a Size

Change View
1 MG
€208.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.46

€208.00


Estimated to ship on30 April 2025Details


Request a Bulk Order

biological source

goat

Quality Level

conjugate

TRITC conjugate
rhodamine conjugate

antibody form

affinity purified immunoglobulin

antibody product type

secondary antibodies

clone

polyclonal

species reactivity

mouse

manufacturer/tradename

Chemicon®

technique(s)

ELISA: suitable
immunofluorescence: suitable

shipped in

wet ice

target post-translational modification

unmodified

Related Categories

General description

IgG is one of the most abundant protein in human serum. IgG is important for our defence against microorganisms and the molecules are produced by B-lymphocytes as a part of our adaptive immune response. The IgG molecule has two separate functions; to bind to the pathogen that elicited the response and to recruit other cells and molecules to destroy the antigen. The variability of the IgG pool is generated by somatic recombination and the number of specificities in an individual at a given time point is estimated to be 1011 variants.

Specificity

Reacts with heavy chain of mouse IgG1, IgG2a, IgG2b and IgG3. Absorbed for mouse IgM and IgA, pooled human sera and purified human paraproteins. Minimal cross reactivity with human immunoglobulins.
The antibody reacts with the heavy chain of mouse IgG1, IgG2a, IgG2b and IgG3 as demonstrated by ELISA and flow cytometry. Minimal cross reactivity with human immunoglobulins.

Immunogen

Epitope: IgG γ chain
Prepared from purified mouse IgG γ chain. Pooled antisera from goats hyperimmunized with mouse IgG paraproteins.

Application

Goat anti-Mouse IgG γ chain Antibody, TRITC conjugate, Species Adsorbed detects level of Mouse IgG γ chain & has been published & validated for use in ELISA & IF.
Immunofluorescence: ≤1 μg/106 cells.
ELISA

Optimal working dilutions must be determined by the end user.
Research Category
Secondary & Control Antibodies
Research Sub Category
Secondary Antibodies Adsorbed for Dual Labeling

Fragment Specific Secondary Antibodies

Physical form

Affinity Chromatography
Purified by affinity chromatography on pooled mouse IgG covalently linked to agarose. Liquid in 1.0 mL PBS/NaN3.

Storage and Stability

Maintain refrigerated at 2°-8°C under sterile conditions for up to twelve months from date of receipt. do not freeze.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Guangchen Ji et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 37(6), 1378-1393 (2016-12-25)
Neuroplasticity in the amygdala drives pain-related behaviors. The central nucleus (CeA) serves major amygdala output functions and can generate emotional-affective behaviors and modulate nocifensive responses. The CeA receives excitatory and inhibitory inputs from the basolateral nucleus (BLA) and serotonin receptor
Guangchen Ji et al.
Molecular brain, 5, 36-36 (2012-10-10)
The medial prefrontal cortex (mPFC) serves major executive functions. mPFC output to subcortical brain areas such as the amygdala controls emotional processing and plays an important role in fear extinction. Impaired mPFC function correlates with extinction deficits in anxiety disorders

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service