Skip to Content
Merck
All Photos(1)

Key Documents

ABS2231

Sigma-Aldrich

Anti-phospho RyR2 (Ser2808)

Synonym(s):

RYR-2, RyR2, hRYR-2, Cardiac muscle ryanodine receptor, Cardiac muscle ryanodine receptor-calcium release channel, Type 2 ryanodine receptor

Sign Into View Organizational & Contract Pricing

Select a Size

25 μG
€137.00

€137.00


Estimated to ship on07 April 2025Details


Request a Bulk Order

Select a Size

Change View
25 μG
€137.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

€137.00


Estimated to ship on07 April 2025Details


Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

mol wt

calculated mol wt 564.57 kDa
observed mol wt ~565 kDa

purified by

affinity chromatography

species reactivity

rabbit

species reactivity (predicted by homology)

mouse, human

packaging

antibody small pack of 100 μg

technique(s)

western blot: suitable

isotype

IgG

epitope sequence

C-terminal cytoplasmic domain

Protein ID accession no.

UniProt accession no.

storage temp.

-10 to -25°C

target post-translational modification

phosphorylation (pSer2808)

Gene Information

human ... RYR2(55193)

General description

Ryanodine receptor 2 (UniProt: Q92736; also known as RYR-2, RyR2, hRYR-2, Cardiac muscle ryanodine receptor, Cardiac muscle ryanodine receptor-calcium release channel, Type 2 ryanodine receptor) is encoded by the RYR2 gene (Gene ID: 6262) in human. Ryanodine receptors are large ion channels that are located in the sarcoplasmic/endoplasmic reticulum membrane and are responsible for the release of Ca2+. They form homotetramers with a mushroom-like shape, consisting of a large cytoplasmic head and transmembrane stalk. In mammals three isoforms of RYR have been described (RYR 1-3) that are regulated by phosphorylation, redox modifications, and a variety of small proteins, and ions. Ryanodine receptor 2 is a multi-pass membrane protein with two cytoplasmic domains (aa 1-4281 and 4871-4967), six transmembrane domains, and one pore-forming domain (aa 4820-4829). It is the predominant form in cardiac muscle and is also expressed in Purkinje cells of cerebellum and cerebral cortex. It plays a key role in triggering cardiac muscle contraction and aberrant channel activation can lead to cardiac arrhythmia. Channel activity is modulated by ryanodine that binds to the open Ca2+-release channel with high affinity. At low concentrations, ryanodine maintains the channel in an open conformation, but high concentrations of ryanodine inhibit channel activity. Channel activity is also modulated by phosphorylation. Phosphorylation at serine 2808 and serine 2814 is shown to increase the open probability of the calcium channel. Phosphorylation is increased in failing heart, leading to calcium leaks, and increased cytoplasmic Ca2+ levels. (Ref.: Xiao, Z., et al. (2016). J. Biol. Chem. 291(47); 24528-24537; Lanner, JT., et al. (2010). Cold Spring Harb. Perspect. Biol. 2(11); a003996).

Specificity

This rabbit polyclonal antibody detects Ryanodine receptor 2 (RyR2) phosphorylated on serine 2808.

Immunogen

A KLH-conjugated linear peptide corresponding to 8 amino acids surrounding phosphoserine 2808 from human Ryanodine receptor 2.

Application

Quality Control Testing

Evaluated by Western Blotting in rabbit skeleton muscle microsomal preparation.

Western Blotting Analysis (WB): A 1:300 dilution of this antibody detected RyR2 phosphorylated on serine 2808 in rabbit skeleton muscle microsomal preparations, but not in microsomes treated with protein phosphatase.

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user

Physical form

Purified rabbit polyclonal antibody in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl without azide

Reconstitution

1.0 mg/mL. Please refer to guidance on suggested starting dilutions and/or titers per application and sample type.

Storage and Stability

Store at -10°C to -25°C. Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service