Skip to Content
Merck
All Photos(2)

Documents

ABN32

Sigma-Aldrich

Anti-PDE9A Antibody

from rabbit

Synonym(s):

High affinity cGMP-specific 3′,5′-cyclic phosphodiesterase 9A

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

polyclonal

species reactivity

human, rhesus macaque, mouse

species reactivity (predicted by homology)

chimpanzee (based on 100% sequence homology), rat (based on 100% sequence homology)

technique(s)

immunohistochemistry: suitable (paraffin)
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... PDE9A(5152)

General description

PDE9A is a ubiquitous protein belonging to the cyclic nucleotide phosphodiesterase family which regulate the signaling of cyclic nucleotides such as cGMP and cAMP. PDE9A has a higher affinity for cGMP. It contains a catalytic site that associates with cofactors and hydrolyzes cGMP thereby terminating cGMP signaling for a number of cellular processes. PDE9A has been indirectly linked to signaling pathways involved in cognitive processes. Previous studies suggested that inhibition of PDE9A enhanced long-term potentiation (a widely studied mechanism of learning and memory); PDE9A may also effect changes in the plasticity of hippocampal neurons which are also important for certain types of cognitive processes. In addition to the brain, PDE9A is also highly expressed in the spleen, prostate, colon, kidney, and heart and may be involved in a range of physiological processes in these systems, via cGMP-amplified signaling.

Immunogen

KLH-conjugated linear peptide corresponding to human PDE9A.

Application

Detect PDE9A using this Anti-PDE9A Antibody validated for use in Western Blotting, IHC(P).
Research Category
Neuroscience
Research Sub Category
Signaling Neuroscience
Western Blot Analysis: A representative lot of this antibody detected PDE9A in human liver tissue lysate, and in mouse spleen and liver tissue lysates.

Immunohistochemistry Analysis: A 1:100 dilution of this antibody detected PDE9 in human prostate tissue, and in possible golgi type II and basket cells in the Purkinje and molecular layers of mouse cerebellum.

Quality

Evaluated by Western Blot in human spleen tissue lysate.

Western Blot Analysis: 0.5 µg/mL of this antibody detected PDE9A in 10 µg of human spleen tissue lysate.

Target description

~75 kDa observed. Uniprot describes 16 isoforms produced by alternative splicing, which have been observed at various molecular weights. (Kruse, L.S., et al. (2009). Brain Res. 1281: 25-34; Rentero, C., et al. (2003). Biochem Biophys Res Commun. 301(3):686-692.)

Physical form

Format: Purified
Protein A purified
Purified rabbit polyclonal in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
Human spleen tissue lysate

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dong I Lee et al.
Nature, 519(7544), 472-476 (2015-03-25)
Cyclic guanosine monophosphate (cGMP) is a second messenger molecule that transduces nitric-oxide- and natriuretic-peptide-coupled signalling, stimulating phosphorylation changes by protein kinase G. Enhancing cGMP synthesis or blocking its degradation by phosphodiesterase type 5A (PDE5A) protects against cardiovascular disease. However, cGMP
Elisa Landucci et al.
ACS chemical neuroscience, 14(20), 3826-3838 (2023-09-20)
In the central nervous system, some specific phosphodiesterase (PDE) isoforms modulate pathways involved in neuronal plasticity. Accumulating evidence suggests that PDE9 may be a promising therapeutic target for neurodegenerative diseases. In the current study, computational techniques were used to identify

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service