Skip to Content
Merck
All Photos(1)

Key Documents

ABN160

Sigma-Aldrich

Anti-ARMS2 Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

Age-related maculopathy susceptibility protein 2

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€351.00

€351.00


Estimated to ship on23 April 2025Details


Request a Bulk Order

Select a Size

Change View
100 μG
€351.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€351.00


Estimated to ship on23 April 2025Details


Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... ARMS2(387715)

General description

Age-related maculopathy susceptibility protein 2 (ARMS2) is highly expressed in the human placenta and retina. The biological function of ARMS2 has not been fully characterized. However, previous studies have linked mutations in the ARMS2 and HtrA1 genes, located on chromosome 10q26, to the occurrence of age-related macular degeneration type 8, a condition of irreversible vision loss in the elderly.

Immunogen

KLH-conjugated linear peptide corresponding to human ARMS2.

Application

Immunocytochemistry Analysis: A representative lot was used by an independent laboratory in ARPE-19 cells. (Pericak-Vance, M., et al. (2009). Invest Ophthalmol Vis Sci. 50(7): 3084–3090.)
Research Category
Neuroscience
Research Sub Category
Neurodegenerative Diseases
This Anti-ARMS2 Antibody is validated for use in Western Blotting for the detection of ARMS2.

Quality

Evaluated by Western Blot in human placenta tissue lysate.

Western Blot Analysis: 1 µg/mL of this antibody detected ARMS2 on 10 µg of human placenta tissue lysate.

Target description

~12 kDa observed. An uncharacterized band appears at ~60 kDa in some lysates.

Physical form

Affinity purified
Purified rabbit polyclonal in PBS containing 0.1% azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
Human placenta tissue lysate

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mao Nakayama et al.
Investigative ophthalmology & visual science, 55(10), 6514-6523 (2014-09-11)
We determined the function of ARMS2 and HtrA1 in the choroid and retina using transgenic (Tg) mice and evaluated the effects of mainstream cigarette smoke on these mice. The chicken actin promoter (CAG) was used to drive mouse HtrA1, human

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service