Skip to Content
Merck
All Photos(2)

Key Documents

ABN1389

Sigma-Aldrich

Anti-MARCO

from rabbit, purified by affinity chromatography

Synonym(s):

Macrophage receptor with collagenous structure, Scavenger receptor class A member 2

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
€326.00

€326.00


Estimated to ship on04 June 2025Details


Request a Bulk Order

Select a Size

Change View
100 μL
€326.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€326.00


Estimated to ship on04 June 2025Details


Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human, mouse

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

ambient

target post-translational modification

unmodified

Gene Information

human ... MARCO(8685)

General description

Macrophage receptor MARCO (UniProt: Q9UEW3; also known as macrophage receptor with collagenous structure; Scavenger receptor class A member 2) is encoded by the MARCO (also known as SCARA2) gene (Gene ID: 8685) in human. MARCO is strongly expressed in macrophages of several tissues in humans with sepsis. It is a scavenger receptor that is readily upregulated following bacterial infection. Hence, it forms a back-up system through which the organism can quickly arm more cells with highly efficient phagocytosing receptors. MARCO is a single-pass type II membrane protein that binds both Gram-positive and Gram-negative bacteria via an extracellular, C-terminal, scavenger receptor cysteine-rich (SRCR) domain. The bacteria-binding region is localized immediately proximal to the cysteine-rich region of domain V. Human and mouse MARCO exhibit about 68% sequence identity. However, the human MARCO subunit chain differs from the mouse chain in that it does not have any cysteine residues outside the cysteine-rich domain V, whereas the mouse chain has one cysteine residue within the intracellular domain I and two in the extracellular spacer domain III. (Ref.: Elomaa, O., et al. (1998). J. Biol. Chem. 273(8): 4350-4538).

Specificity

This polyclonal antibody detects macrophage receptor MARCO in RAW264.7 cells.

Immunogen

KLH-cojugated peptide corresponding to 12 amino acids from the C-terminal region of human MARCO.

Application

Anti-MARCO, Cat. No. ABN1389, is a highly specific rabbit polyclonal antibody that targets Macrophage receptor MARCO and has been tested in Western Blotting.
Research Category
Neuroscience
Western Blotting Analysis: A 1:500 dilution from a representative lot detected MARCO in 10 µg of A549 cell lysate.

Quality

Evaluated by Western Blotting in RAW264.7 cells.

Western Blotting Analysis (WB): A 1:5000 Dilution of this antibody detected Macrophage receptor MARCO in 10 µg RAW264.7 cells .

Target description

~60 kDa observed; 52.65 kDa calculated. Uncharacterized bands may be observed in some lysate(s).

Physical form

Affinity Purified
Purified rabbit polyclonal antibody in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service