Skip to Content
Merck
All Photos(1)

Key Documents

ABE1961

Sigma-Aldrich

Anti-Borealin

serum, from rabbit

Synonym(s):

Cell division cycle-associated protein 8, Dasra-B, hDasra-B, Pluripotent embryonic stem cell-related gene 3 protein

Sign Into View Organizational & Contract Pricing

Select a Size

50 μL
€397.00

€397.00


In StockDetails


Request a Bulk Order

Select a Size

Change View
50 μL
€397.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.43

€397.00


In StockDetails


Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

serum

antibody product type

primary antibodies

clone

polyclonal

species reactivity

human

technique(s)

immunofluorescence: suitable
immunoprecipitation (IP): suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

ambient

target post-translational modification

unmodified

Gene Information

human ... CDCA8(55143)

General description

Borealin (UniProt: Q53HL2; also known as Cell division cycle-associated protein 8; Dasra-B; hDasra-B; Pluripotent embryonic stem cell-related gene 3 protein) is encoded by the CDCA8 (also known as PESCRG3) gene (Gene ID: 55143) in human. Borealin is a novel 31-kDa chromosomal passenger protein that is conserved among all vertebrates. It is a member of the chromosomal passenger holocomplex along with Aurora B, INCENP, and survivin. This complex play a vital role at the centromere to ensure proper chromosome alignment and segregation. Borealin is required for stability of the bipolar mitotic spindle and for kinetochore error correction. Borealin levels are reported to increase during G2/M phase and are reduced following exit from the M phase of cell cycle. Depletion of Borealin by RNAi is shown to retard mitotic progression and cause spindle T misattachment and abnormalities. Borealin-depleted cells are shown to align their chromosomes to a near normal metaphase plate, but they abruptly undergo multipolar anaphases.

Specificity

This polyclonal antibody detects Borealin in nocodozole treated HeLa cells.

Immunogen

A His-tagged full length human recombinant Borealin.

Application

Anti-Borealin Antibody, Cat. No. ABE1961, is a highly specific rabbit polyclonal antibody that targets Borealin and has been tested in Immunofluorescence, Immunoprecipitation, and Western Blotting.
Immunofluorescence Analysis: A representative lot detected Borealin in HeLa cells (Gassmann, R., et. al. (2004). J Cell Biol. 166(2):179-91).

Western Blotting Analysis: A representative lot detected Borealin in HeLa cells (Gassmann, R., et. al. (2004). J Cell Biol. 166(2):179-91).

Immunoprecipitation Analysis: A representative lot detected Borealin in HeLa cells (Gassmann, R., et. al. (2004). J Cell Biol. 166(2):179-91).

Quality

Evaluated by Western Blotting in Hela cell lysate treated with Nocodozole.

Western Blotting Analysis: A 1:5,000 dilution of this antibody detected Borealin in 10 µg of Hela cell lysate treated with Nocodozole.

Target description

~36 kDa observed; 31.32 kDa calculated. Uncharacterized bands may be observed in some lysate(s).

Other Notes

Concentration: Please refer to lot specific datasheet.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service