Skip to Content
Merck
All Photos(2)

Documents

ABE1361

Sigma-Aldrich

Anti-NuMA1 Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

Nuclear mitotic apparatus protein 1, NuMA protein, SP-H antigen

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human

technique(s)

immunocytochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... NUMA1(4926)

General description

NuMA1 or NuMA protein is an interesting and major component of the nuclear matrix of eukaryotic cells. NuMA1 is a highly abundant protein component that appears to serve a non-structural role but is needed for mitotic spindle poles and centrosome formation as well as a linker of microtubules. These functions are important for proper cytokinesis and the setup of the proper polarity axis during cell division. NuMA1 resides in the nuclear matrix during interphase. NuMA1 dissociates from condensing chromosomes during early prophase, and relocates to the spindle poles via a dynein/dynamin complex, and it remains there until the anaphase onset. Thus NuMA1 then can be found in the nucleus during cell rest and cytoplasmically and centrosome localized during cytokinesis.

Specificity

Recognizes NuMA1

Immunogen

Three different peptides corresponding to multiple locations of NuMA1 isoform 1. Peptide, region between amino acids 1-240. Peptide 2, amino acids 1300-1500. Peptide 3, amino acids 1860-2115. Note that these peptides are also found in isoform 2.

Application

Immunocytochemistry Analysis: A 1:100 dilution from a representative lot was reported to detect NuMA1 in HeLa cells.
Research Category
Epigenetics & Nuclear Function

Epigenetics & Nuclear Function
Research Sub Category
Cell Cycle, DNA Replication & Repair

Chromatin Biology
This Anti-NuMA1 Antibody is validated for use in Western Blotting and Immunocytochemistry for the detection of NuMA1.

Quality

Evaluated by Western Blotting in A431 cell lysate.

Western Blotting Analysis: A 1 µg/mL dilution of this antibody detected NuMA1 in a A431 cell lysate.

Target description

~ 238 kDa observed

Physical form

Antigen Affinity Purified
Purified rabbit polyclonal in buffer containing PBS, 50% glycerol, and 0.05% sodium azide.

Storage and Stability

Store at -20°C

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

K S Aboody et al.
Proceedings of the National Academy of Sciences of the United States of America, 97(23), 12846-12851 (2000-11-09)
One of the impediments to the treatment of brain tumors (e.g., gliomas) has been the degree to which they expand, infiltrate surrounding tissue, and migrate widely into normal brain, usually rendering them "elusive" to effective resection, irradiation, chemotherapy, or gene
Anna Rodina et al.
Nature communications, 14(1), 3742-3742 (2023-06-24)
Systems-level assessments of protein-protein interaction (PPI) network dysfunctions are currently out-of-reach because approaches enabling proteome-wide identification, analysis, and modulation of context-specific PPI changes in native (unengineered) cells and tissues are lacking. Herein, we take advantage of chemical binders of maladaptive

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service