Skip to Content
Merck
All Photos(2)

Documents

AB9714-I

Sigma-Aldrich

Anti-RAGE

from rabbit, purified by affinity chromatography

Synonym(s):

Advanced glycosylation end product-specific receptor, Receptor for advanced glycosylation end products, RAGE

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.43

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

mouse, rat

species reactivity (predicted by homology)

human (based on 100% sequence homology)

technique(s)

immunohistochemistry: suitable (paraffin)
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

ambient

target post-translational modification

glycosylation

Gene Information

human ... AGER(177)

General description

Advanced glycosylation end product-specific receptor (UniProt: Q15109; also known as Receptor for advanced glycosylation end products, RAGE) is encoded by the AGER (also known as RAGE) gene (Gene ID: 177) in human. RAGE is a single-pass type I membrane protein that contains two Ig-like C2-type domains, one Ig-like V-type domain, a 21 amino acid transmembrane domain and a 41 amino acids cytoplasmic domain. RAGE mediates interactions of advanced glycosylation end products (AGE), which are non-enzymatically glycosylated proteins which accumulate in vascular tissue in aging and at an accelerated rate in diabetes. RAGE acts as a mediator of both acute and chronic vascular inflammation in conditions, such as atherosclerosis and in complication of diabetes. Blockage of RAGE is reported to suppress development of atherosclerosis and vascular disorders. RAGE is expressed by endothelium, mononuclear phagocytes, neurons and smooth muscle cells. Higher expression of RAGE is observed during development, especially in the central nervous system, and its levels decline during maturity. Besides binding to advanced glycosylation end product (AGE), RAGE also binds amyloid beta peptide, S100family proteins, and leukocyte integrins. Excessive amounts of amyloid peptides (Ab) in brain leads to the hyperstimulation of RAGE. The RAGE-Ab interaction is thought to result in oxidative stress leading to neuronal degeneration.

Specificity

This polyclonal antibody detects Receptor for Advanced Glycosylation End Products (RAGE). This antibody targets an epitope corresponding to 21 amino acids in the N-terminal region of human RAGE. Human, mouse, and rat RAGE display 95% homology in this epitope region.

Immunogen

KLH-cojugated linear peptide corresponding to 20 amino acids from the N-terminal region of human RAGE.

Application

Anti-RAGE, Cat. No. AB9714-I, is a highly specific rabbit polyclonal antibody that targets Advanced glycosylation end product-specific receptor and has been tested in Immunohistochemistry (Paraffin) and Western Blotting.
Immunohistochemistry Analysis: A 1:1,000 dilution from a representative lot detected RAGE in human lung tissue.
Research Category
Neuroscience

Quality

Evaluated by Western Blotting in mouse lung tissue lysate.

Western Blotting Analysis: 2 µg/mL of this antibody detected RAGE in 10 µg of mouse lung tissue lysate.


Target description

~48 kDa observed; 42.80 kDa calculated. Uncharacterized bands may be observed in some lysate(s).

Linkage

Replaces: AB9714

Physical form

Affinity Purified
Purified rabbit polyclonal antibody in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service